CO1H324A6FD6 CombiPhos Catalysts Inc

2,4-Dimethyl-5-cyclopropylphenylboronic acid pinacol ester

  • Cas Number : 2223054-21-3
  • Molecular Weight : 272.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin US
Availability details for
Quantity
  • Availability
  • Add to Cart