CO1H324A6FD8 CombiPhos Catalysts Inc

2-Methyl-3-trifluoromethyl-5-cyclopropylphenylboronic acid pinacol ester

  • Cas Number : 2223034-87-3
  • Molecular Weight : 326.17

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin US
Availability details for
Quantity
  • Availability
  • Add to Cart