CO1H324A6FDD CombiPhos Catalysts Inc

2-Methyl-4-methoxy-5-cyclopropylphenylboronic acid pinacol ester

  • Cas Number : 2223041-86-7
  • Molecular Weight : 288.19

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin US
Availability details for
Quantity
  • Availability
  • Add to Cart