CO1H324A6FDE CombiPhos Catalysts Inc

2-Chloro-3-cyano-5-cyclopropylphenylboronic acid pinacol ester

  • Cas Number : 2223033-61-0
  • Molecular Weight : 303.6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin US
Availability details for
Quantity
  • Availability
  • Add to Cart