CO1H324A6FE0 CombiPhos Catalysts Inc

2-Chloro-4-methoxy-5-cyclopropylphenylboronic acid pinacol ester

  • Cas Number : 2223046-41-9
  • Molecular Weight : 308.61

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin US
Availability details for
Quantity
  • Availability
  • Add to Cart