CO1H324A6FE3 CombiPhos Catalysts Inc

2-Fluoro-3-methyl-5-cyclopropylphenylboronic acid pinacol ester

  • Cas Number : 2223037-36-1
  • Molecular Weight : 276.16

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin US
Availability details for
Quantity
  • Availability
  • Add to Cart