CO1H324A6FE4 CombiPhos Catalysts Inc

3-Chloro-6-methyl-4-cyclopropylphenylboronic acid pinacol ester

  • Cas Number : 2223031-72-7
  • Molecular Weight : 292.61

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin US
Availability details for
Quantity
  • Availability
  • Add to Cart