CO1H324A6FE9 CombiPhos Catalysts Inc

3-Bromo-5-cyclopropyl-6-methylphenylboronic acid pinacol ester

  • Cas Number : 2223042-71-3
  • Molecular Weight : 337.06

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin US
Availability details for
Quantity
  • Availability
  • Add to Cart