CO1H324A6FEA CombiPhos Catalysts Inc

2-Chloro-5-trifluoromethyl-4-methylphenylboronic acid pinacol ester

  • Cas Number : 2223041-88-9
  • Molecular Weight : 320.55

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin US
Availability details for
Quantity
  • Availability
  • Add to Cart