CO1H324A6FEB CombiPhos Catalysts Inc

2-Chloro-5-trifluoromethyl-4-cyclopropylphenylboronic acid pinacol ester

  • Cas Number : 2223033-71-2
  • Molecular Weight : 346.58

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin US
Availability details for
Quantity
  • Availability
  • Add to Cart