CO1H324A6FED CombiPhos Catalysts Inc

2-Methyl-5-cyano-3-cyclopropylphenylboronic acid pinacol ester

  • Cas Number : 2223040-47-7
  • Molecular Weight : 283.18

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin US
Availability details for
Quantity
  • Availability
  • Add to Cart