CO1H324A6FF0 CombiPhos Catalysts Inc

2-Chloro-4-methyl-5-methoxyphenylboronic acid pinacol ester

  • Cas Number : 2223031-75-0
  • Molecular Weight : 282.57

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin US
Availability details for
Quantity
  • Availability
  • Add to Cart