CO1H324A6FF5 CombiPhos Catalysts Inc

2-Fluoro-4-methyl-5-cyclopropylphenylboronic acid pinacol ester

  • Cas Number : 2223042-83-7
  • Molecular Weight : 276.16

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin US
Availability details for
Quantity
  • Availability
  • Add to Cart