CO1H324A6FF8 CombiPhos Catalysts Inc

2-Bromo-3-methyl-5-trifluoromethylphenylboronic acid pinacol ester

  • Cas Number : 2223033-86-9
  • Molecular Weight : 365

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin US
Availability details for
Quantity
  • Availability
  • Add to Cart