CO1H324A6FF9 CombiPhos Catalysts Inc

2-Bromo-3-cyclopropyl-5-trifluoromethylphenylboronic acid pinacol ester

  • Cas Number : 2223055-31-8
  • Molecular Weight : 391.04

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin US
Availability details for
Quantity
  • Availability
  • Add to Cart