CO1H324A6FFB CombiPhos Catalysts Inc

3-Cyclopropyl-2,5-dimethylphenylboronic acid pinacol ester

  • Cas Number : 2223047-64-9
  • Molecular Weight : 272.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin US
Availability details for
Quantity
  • Availability
  • Add to Cart