CO1H324A7006 CombiPhos Catalysts Inc

2-Chloro-3-cyclopropyl-5-trifluoromethylphenylboronic acid pinacol ester

  • Cas Number : 2223035-15-0
  • Molecular Weight : 346.58

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin US
Availability details for
Quantity
  • Availability
  • Add to Cart