CO1H324A7007 CombiPhos Catalysts Inc

3-Chloro-4-bromo-6-methylphenylboronic acid pinacol ester

  • Cas Number : 2223046-57-7
  • Molecular Weight : 331.44

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin US
Availability details for
Quantity
  • Availability
  • Add to Cart