CO1H324A700A CombiPhos Catalysts Inc

3-Chloro-4,6-dimethylphenylboronic acid pinacol ester

  • Cas Number : 2223048-51-7
  • Molecular Weight : 266.58

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin US
Availability details for
Quantity
  • Availability
  • Add to Cart