CO1H324A700D CombiPhos Catalysts Inc

2-Chloro-3-methyl-5-cyanophenylboronic acid pinacol ester

  • Cas Number : 2223033-38-1
  • Molecular Weight : 277.56

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin US
Availability details for
Quantity
  • Availability
  • Add to Cart