CO1H324A7012 CombiPhos Catalysts Inc

4-Chloro-5-trifluoromethyl-2-cyclopropylphenylboronic acid pinacol ester

  • Cas Number : 2223035-32-1
  • Molecular Weight : 346.58

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin US
Availability details for
Quantity
  • Availability
  • Add to Cart