CO1H324A7016 CombiPhos Catalysts Inc

2,5-Dimethyl-4-cyclopropylphenylboronic acid pinacol ester

  • Cas Number : 2223046-62-4
  • Molecular Weight : 272.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin US
Availability details for
Quantity
  • Availability
  • Add to Cart