CO1H324A7023 CombiPhos Catalysts Inc

2-(3-cyclopropyl-5-isopropylphenyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane

  • Cas Number : 2223040-66-0
  • Molecular Weight : 286.22

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin US
Availability details for
Quantity
  • Availability
  • Add to Cart