CO1H324A7024 CombiPhos Catalysts Inc

3,5-Dicyclopropylphenylboronic acid pinacol ester

  • Cas Number : 1202644-27-6
  • Molecular Weight : 284.21

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin US
Availability details for
Quantity
  • Availability
  • Add to Cart