CO1H324A7025 CombiPhos Catalysts Inc

3-(Cyclopropyl)-5-(cyclobutyl)phenylboronic acid pinacol ester

  • Cas Number : 2223033-63-2
  • Molecular Weight : 298.23

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin US
Availability details for
Quantity
  • Availability
  • Add to Cart