CO1H324A702C CombiPhos Catalysts Inc

3-Methyl-5-(2-tetrahydrofuranyl)phenylboronic acid pinacol ester

  • Cas Number : 2223004-56-4
  • Molecular Weight : 288.19

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin US
Availability details for
Quantity
  • Availability
  • Add to Cart