CO1H324A702E CombiPhos Catalysts Inc

3-(3-tetrahydrofuranyl)-5-trifluoromethylphenylboronic acid pinacol ester

  • Cas Number : 2222996-59-8
  • Molecular Weight : 342.17

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin US
Availability details for
Quantity
  • Availability
  • Add to Cart