CO1H324A7036 CombiPhos Catalysts Inc

4-(Thiazol-5-yl)-2-fluorophenylboronic acid pinacol ester

  • Cas Number : 2223043-18-1
  • Molecular Weight : 305.18

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin US
Availability details for
Quantity
  • Availability
  • Add to Cart