CO1H324A703B CombiPhos Catalysts Inc

3-Cyclopropyl-5-(thiazol-4-yl)phenylboronic acid pinacol ester

  • Cas Number : 2223031-91-0
  • Molecular Weight : 327.26

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin US
Availability details for
Quantity
  • Availability
  • Add to Cart