CO1H324A703E CombiPhos Catalysts Inc

3-Bromo-5-(thiazol-5-yl)phenylboronic acid pinacol ester

  • Cas Number : 2223048-66-4
  • Molecular Weight : 366.09

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin US
Availability details for
Quantity
  • Availability
  • Add to Cart