CO1H324A7041 CombiPhos Catalysts Inc

2-(N-Boc-piperidin-4-yl)phenylboronic acid pinacol ester

  • Cas Number : 2223051-40-7
  • Molecular Weight : 387.33

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin US
Availability details for
Quantity
  • Availability
  • Add to Cart