CO1H324A7047 CombiPhos Catalysts Inc

3-Amino-5-(N-Boc-piperidin-4-yl)phenylboronic acid pinacol ester

  • Cas Number : 2223041-13-0
  • Molecular Weight : 402.34

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin US
Availability details for
Quantity
  • Availability
  • Add to Cart