CO1H324A704E CombiPhos Catalysts Inc

3-(4-Tetrahydropyranyl)-5-(N-Boc-piperidin-4-yl)phenylboronic acid pinacol ester

  • Cas Number : 2223051-46-3
  • Molecular Weight : 471.45

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin US
Availability details for
Quantity
  • Availability
  • Add to Cart