CO1H324A7053 CombiPhos Catalysts Inc

4-(2-Fluorophenyl)thiophene-2-boronic acid pinacol ester

  • Cas Number : 1402174-27-9
  • Molecular Weight : 304.19

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin US
Availability details for
Quantity
  • Availability
  • Add to Cart