CO1H324A7058 CombiPhos Catalysts Inc

3-Chloro-4-(N-Boc-piperidin-3-yl)phenylboronic acid pinacol ester

  • Cas Number : 2222996-45-2
  • Molecular Weight : 421.77

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin US
Availability details for
Quantity
  • Availability
  • Add to Cart