CO1H324A705B CombiPhos Catalysts Inc

3-Trifluoromethyl-4-(N-Boc-piperidin-3-yl)phenylboronic acid pinacol ester

  • Cas Number : 2222996-61-2
  • Molecular Weight : 455.33

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin US
Availability details for
Quantity
  • Availability
  • Add to Cart