CO1H324A705D CombiPhos Catalysts Inc

2-Fluoro-5-(N-Boc-piperidin-3-yl)phenylboronic acid pinacol ester

  • Cas Number : 2223004-63-3
  • Molecular Weight : 405.32

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin US
Availability details for
Quantity
  • Availability
  • Add to Cart