CO1H324A7065 CombiPhos Catalysts Inc

2-(N-Boc-Piperidin-3-yl)-4-methylphenylboronic acid pinacol ester

  • Cas Number : 2223009-78-5
  • Molecular Weight : 401.35

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin US
Availability details for
Quantity
  • Availability
  • Add to Cart