CO1H324A7067 CombiPhos Catalysts Inc

3-(N-Boc-Piperidin-3-yl)-5-trifluoromethylphenylboronic acid pinacol ester

  • Cas Number : 2222997-17-1
  • Molecular Weight : 455.33

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin US
Availability details for
Quantity
  • Availability
  • Add to Cart