CO1H324A7069 CombiPhos Catalysts Inc

4-(2-Tolyl)furan-2-boronic acid pinacol ester

  • Cas Number : 2096331-39-2
  • Molecular Weight : 284.16

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin US
Availability details for
Quantity
  • Availability
  • Add to Cart