CO1H324A706D CombiPhos Catalysts Inc

3-(N-Boc-Piperidin-3-yl)-5-bromophenylboronic acid pinacol ester

  • Cas Number : 2223009-35-4
  • Molecular Weight : 466.22

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin US
Availability details for
Quantity
  • Availability
  • Add to Cart