CO1H324A706F CombiPhos Catalysts Inc

4-(N-Boc-Piperidin-4-yl)-3-chlorophenylboronic acid pinacol ester

  • Cas Number : 2223048-88-0
  • Molecular Weight : 421.77

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin US
Availability details for
Quantity
  • Availability
  • Add to Cart