CO1H324A7071 CombiPhos Catalysts Inc

4-(N-Boc-Piperidin-4-yl)-3-trifluoromethylphenylboronic acid pinacol ester

  • Cas Number : 2223047-69-4
  • Molecular Weight : 455.33

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin US
Availability details for
Quantity
  • Availability
  • Add to Cart