CO1H324A7073 CombiPhos Catalysts Inc

tert-butyl 4-(2-amino-5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl)piperidine-1-carboxylate

  • Cas Number : 2223043-79-4
  • Molecular Weight : 402.34

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin US
Availability details for
Quantity
  • Availability
  • Add to Cart