CO1H324A7075 CombiPhos Catalysts Inc

3-(N-Boc-Piperidin-4-yl)-4-fluorophenylboronic acid pinacol ester

  • Cas Number : 2223035-72-9
  • Molecular Weight : 405.32

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin US
Availability details for
Quantity
  • Availability
  • Add to Cart