CO1H324A7078 CombiPhos Catalysts Inc

3-(N-Boc-Piperidin-4-yl)-4-methylphenylboronic acid pinacol ester

  • Cas Number : 2223051-66-7
  • Molecular Weight : 401.35

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin US
Availability details for
Quantity
  • Availability
  • Add to Cart