CO1H324A707C CombiPhos Catalysts Inc

5-(N-Boc-Piperidin-4-yl)-2-methoxyphenylboronic acid pinacol ester

  • Cas Number : 2223041-96-9
  • Molecular Weight : 417.35

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin US
Availability details for
Quantity
  • Availability
  • Add to Cart