CO1H324A707D CombiPhos Catalysts Inc

5-(N-Boc-Piperidin-4-yl)-2-cyanophenylboronic acid pinacol ester

  • Cas Number : 2223047-71-8
  • Molecular Weight : 412.34

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin US
Availability details for
Quantity
  • Availability
  • Add to Cart