CO1H324A707F CombiPhos Catalysts Inc

6-Methyl-2-chloropyrimidine-4-boronic acid pinacol ester

  • Cas Number : 1310405-15-2
  • Molecular Weight : 254.52

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin US
Availability details for
Quantity
  • Availability
  • Add to Cart