CO1H324A7081 CombiPhos Catalysts Inc

3-(SEC-BUTYL)PHENYLBORONIC ACID PINACOL ESTER

  • Cas Number : 2223009-79-6
  • Molecular Weight : 260.18

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin US
Availability details for
Quantity
  • Availability
  • Add to Cart